Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD IMag™ Buffer

Catalog No. BDB552362 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552362 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552362 Supplier BD Supplier No. 552362
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the optimal labeling of leukocytes using BD IMag particles and for optimal magnetic cell separation using the BD IMagnet™ Cell Separation Magnet

A 10X solution containing bovine serum albumin, EDTA, and sodium azide in phosphate buffered saline (PBS).

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Content And Storage 4°C
For Use With (Application) Magnetic Cell Separation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.