Learn More
BD Interlink™ Vial Access Cannulas
Catalog No. p-3239162 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Volume (Metric):
3 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-689-7C 5 mL
13-689-7A 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-689-7C Supplier BD Supplier No. 303403
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Interlink System syringes with cannula used for needleless access of a single-dose vial
  • Specifically designed for use with Interlink injection sites
  • Identified by a colored alert ring around the septum
  • BD safety-engineered device
  • Latex-free
Not compatible with conventional injection sites.
IV flush, IV push
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 15 G
For Use With (Application) For needleless access of a single dose vial, IV flush, IV push, for use with Baxter™ Interlink™, Abbott Lifeshield, McGaw Safeline™
Product Type Syringe
Volume (Metric) 5 mL
Sterility Sterile