Learn More
Description
- Source is rich in endogenously expressed proteins
- Provided in denatured format allowing for immediate loading onto SDS-PAGE gel
- Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
- Concentration: 1mg/mL
Routinely tested western blot
Specifications
Specifications
| Quantity | 500 μg |
| Storage Buffer | 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol |
| Host Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
| For Use With (Application) | Western Blot |
| Tissue | T-cell Leukemia |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.