Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Jurkat + Pervanadate Lysate

Catalog No. BDB611755 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611755 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611755 Supplier BD Supplier No. 611755

missing translation for 'validMainframeMsg'

Includes

Human T-Cell Leukemia

For blotting procedures

  • Concentration: 1.0mg/mL
  • Rich in endogenously expressed proteins
  • T-Cells were grown to 80-90% confluency

Western blot

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue T-cell Leukemia
Preparation Method Jurkat (T-cell leukemia) cells, were grown to 80 to 90% confluency, then treated with 1mM pervanadate, a general inhibitor of Protein Tyrosine Phosphatases, for 15 min. at 37°C
Quantity 500 μg
Storage Buffer 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Host Species Human
Concentration 1 mg/mL
Content And Storage -20°C

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.