Learn More
BD Macro-Vue™ RPR Cards and Components
Catalog No. 22349688 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1.5 mL
10 Pack
100 Pack
300 Case
300 Pack
500 Tests
Type:
BD RPR Cards
Dispensing Needles, 20 Gauge
Liquid RPR Controls
RPR Antigen Dispensing Bottles
RPR BD Cards
RPR Control Cards
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-349688 500 Tests -
L4972001 100 Pack RPR BD Cards
L4970509 10 Pack RPR Antigen Dispensing Bottles
L4971849 300 Case BD RPR Cards
L4971949 300 Pack RPR BD Cards
22-130989 10 Pack Dispensing Needles, 20 Gauge
L4976909 1.5 mL Liquid RPR Controls
22-727891 10 Pack RPR Control Cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-349688 Supplier BD Supplier No. B275005
This item is not returnable. View return policy
For large-scale testing in public health and clinical laboratories
RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.
  • CLIA Complexity: Moderate
  • Nontreponemal test
  • For the serological detection of syphilis as described in A Manual of Tests for Syphilis
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For large-scale testing in public health and clinical laboratories
Quantity 500 Tests