Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Macro-Vue™ RPR Cards and Components

Catalog No. L4976909 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1.5 mL
10 Pack
100 Pack
300 Case
300 Pack
500 Tests
Type:
BD RPR Cards
Dispensing Needles, 20 Gauge
Liquid RPR Controls
RPR Antigen Dispensing Bottles
RPR BD Cards
RPR Control Cards
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
L4976909 1.5 mL Liquid RPR Controls
L4972001 100 Pack RPR BD Cards
L4970509 10 Pack RPR Antigen Dispensing Bottles
L4971849 300 Case BD RPR Cards
L4971949 300 Pack RPR BD Cards
22-130989 10 Pack Dispensing Needles, 20 Gauge
22-349688 500 Tests -
22-727891 10 Pack RPR Control Cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. L4976909 Supplier BD Supplier No. 276909
This item is not returnable. View return policy

For large-scale testing in public health and clinical laboratories

RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.

  • CLIA Complexity: Moderate
  • Nontreponemal test
  • For the serological detection of syphilis as described in A Manual of Tests for Syphilis

TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Reagin (Syphilis)
Type Liquid RPR Controls
Formulation Contains pooled Human Serum with 0.1% Sodium Azide as a preservative
For Use With (Application) For large-scale testing in public health and clinical laboratories
Includes BD Macro-Vue RPR liquid controls, 1.5mL
Quantity 1.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.