Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD MAX™ System

Catalog No. B441916 Shop All Waters Corporation Products
Change view
Click to view available options
Depth (English):
28.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English)
B441916 28.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B441916 Supplier BD Supplier No. 441916

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fully-integrated, automated molecular platform capable of running both IVD and open system assays efficiently and flexibly

BD MAX System is intended for in vitro diagnostic (IVD) use in performing FDA cleared or approved nucleic acid testing in clinical laboratories. It is capable of automated extraction and purification of nucleic acids from multiple specimen types, as well as the automated amplification and detection of target nucleic acid sequences by fluorescence-based PCR.

The BD MAX System is capable of:

  • Independent nucleic acid extraction and purification
  • PCR amplification using thermal cycling across 24 lanes
  • Real time detection of products of amplification
  • Identification of each Sample Buffer tube by external barcode reader
  • Work List verification by internal barcode reader
  • Fluorescence detection at up to five wavelengths, allowing multiplex reactions by Cycle threshold (Ct) call, or multiplex reactions by melt analysis
  • Qualitative, quantitative and melt sample analysis

Features:

  • Integrated and fully-automated extraction and thermocycling steps
  • Processes and analyzes up to 24 specimens per run
  • On-board lysis and extraction with pre-filled reagent strips
  • Real-time PCR with two 24-lane microfluidic PCR readers
  • Dedicated multi-color optics including: 475/520, 530/565, 585/630, 630/665, 680/715
  • Consistent results through standardized workflow and protocols
  • Rapidly expanding menu and open-system options to help address emerging testing needs
  • Less than one minute of hands on time per sample
  • CLIA Moderately complex and easy to use
  • All-in-One computer contains instrument software that is responsible for instrument control, self-calibration, data analysis and storage
  • User interface can be navigated with point-and-click icons (keyboard and mouse), or the Computer's touchscreen

Modules included:

  • Temperature control/heating subsystem to control thermal cycling
  • A liquid handling head to transfer samples from Sample Buffer tubes to Unitized Reagent Strips
  • Onboard readers to measure the amplification reaction and report results

Software/user interface allows for:

  • 24 unique thermal cycling channels per system (two onboard readers)
  • Entering patient/sample information, either manually or by external barcode reader
  • Monitoring operations, communication and storing of data
  • Qualitative, quantitative and melt sample analysis
  • Error warnings
  • Download of results to a USB thumb drive
  • LIS connectivity

Certifications

CE marked

TRUSTED_SUSTAINABILITY

Specifications

Electrical Requirements 100/240 V, 50/60 Hz, 10 A, 300 VA Max
Height (English) 37 in.
Depth (English) 28.5 in.
Width (English) 29.7 in.
Weight 250 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.