Learn More
BD MAX™ MRSA XT Assay Kit
Catalog No. B443461 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Includes:
BD MAX™ MRSA XT Master Mix, BD MAX™ MRSA XT Reagent Strip, BD MAX™ MRSA XT Extraction Tube, BD MAX™ MRSA XT Sample Buffer Tube
Catalog No. Includes
B443461 BD MAX™ MRSA XT Master Mix, BD MAX™ MRSA XT Reagent Strip, BD MAX™ MRSA XT Extraction Tube, BD MAX™ MRSA XT Sample Buffer Tube
1 options
Catalog No. B443461 Supplier BD Supplier No. 443461
Add to Cart
Add to Cart
Includes
BD MAX MRSA XT Master Mix, BD MAX MRSA XT Reagent Strip, BD MAX MRSA XT Extraction Tube, BD MAX MRSA XT Sample Buffer Tube
Automated qualitative in vitro diagnostic test for detection of methicillin-resistant Staphylococcus aureus (MRSA) DNA from nasal swabs in patients at risk for nasal colonization
The test utilizes real-time polymerase chain reaction (PCR) for the amplification of MRSA DNA and fluorogenic target-specific hybridization probes for the detection of amplified DNA: specific amplicon in the SCCmec right-extremity junction (MREJ), the genes for methicillin resistance mecA and mecC, the gene nuc encoding a thermostable nuclease of S. aureus. The assay is intended to aid in the prevention and control of MRSA infections in healthcare settings.
Performed on BD MAX™ System
TRUSTED_SUSTAINABILITY
Specifications
Includes BD MAX™ MRSA XT Master Mix, BD MAX™ MRSA XT Reagent Strip, BD MAX™ MRSA XT Extraction Tube, BD MAX™ MRSA XT Sample Buffer Tube