Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD MCF7 Cell Lysate

Catalog No. BDB611548 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611548 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611548 Supplier BD Supplier No. 611548

missing translation for 'validMainframeMsg'

Convenient to use and useful control for western blotting application

  • Source is rich in endogenously expressed proteins
  • Provided in denatured format allowing for immediate loading onto SDS-PAGE gel
  • Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
  • Concentration: 1mg/mL

Routinely tested western blot

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue Breast Adenocarcinoma
Quantity 500 μg
Storage Buffer 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Host Species Human
Concentration 1 mg/mL
Content And Storage -20°C

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.