Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Media Supplement Fluid A

Catalog No. B4392391 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
For Use With (Application):
Dehydrated culture media
Sterility Testing
Content And Storage:
15°C to 30°C
2 to 25°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application) Content And Storage
B4392391 100 mL Dehydrated culture media 2 to 25°C
B99111 500 mL Sterility Testing 15°C to 30°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4392391 Supplier BD Supplier No. 292391
Add to Cart
Add to Cart

For diluting and rinsing in sterility testing

  • Used in microbial limit testing, food, dairy and water testing

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2 to 25°C
Additive Fluid A
Form Liquid
For Use With (Application) Dehydrated culture media
Packaging Type Bottle
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.