Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse GM-CSF ELISA Set

Catalog No. BDB555167 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555167 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555167 Supplier BD Supplier No. 555167
Add to Cart
Add to Cart

For mouse granulocyte macrophage colony stimulating factor (GM-CSF)

The OptEIA™ Set contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant mouse GM-CSF in serum, plasma, and cell culture supernatants.

  • Capture antibody: anti-mouse GM-CSF
  • Detection antibody: biotinylated anti-mouse GM-CSF
  • Enzyme reagent: streptavidin-horseradish peroxidase conjugate (SAv-HRP)
  • Standard: lyophilized recombinant mouse GM-CSF
  • Standard range: 15.6 - 1000 pg/mL

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 15.6 to 1000pg/mL
Product Type Mouse GM-CSF ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Mouse GM-CSF in Serum, Plasma and cell culture supernatants
Kit Contents anti-Mouse GM-CSF monoclonal (capture) antibody, Biotinylated anti-Mouse GM-CSF monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse GM-CSF (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.