Learn More
Description
Includes
Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant mouse IL-1β in serum, plasma, and cell culture supernatant
Enzyme-linked immunosorbent assay
Specifications
Specifications
| Product Type | Mouse IL-1β ELISA set |
| Sufficient For | 20 x 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant mouse IL-1βbeta in serum, plasma and cell culture supernatants |
| Kit Contents | anti-Mouse IL-1β monoclonal (capture) antibody, Biotinylated anti-Mouse IL-1β monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IL-1β (standard), Streptavidin-HRP Conjugate |
| Storage Requirements | 2°C to 8°C |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.