Learn More
BD Mouse Th1/Th2/Th17 Phenotyping Kit
Catalog No. BDB560758 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB560758 1 Kit
1 options
Catalog No. BDB560758 Supplier BD Supplier No. 560758
Add to Cart
Add to Cart
Includes
Mouse Th1/Th2/Th17 Phenotyping Cocktails (CD4 PerCP-Cy™5.5, IL-17A PE, IFN- γFITC, IL-4 APC), BD Cytofix™ Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Simultaneously view Th17, Th2 and Th17 cells in mouse model systems
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Can be used to analyze ex vivo lymphoid cell samples or to monitor T helper cell differentiation
  • Ensures indentification and characterization of the nature of the T helper cell types
  • Allow for staining of surface CD4 along with intracellular factors
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For research use only. Not for diagnostic or therapeutic procedures.