Learn More
BD Cell Analysis 3P IMK Kit
Supplier:  BD Cell Analysis 3P 662965
Encompass
The BD Multitest™ IMK kit is a four-color direct immunofluorescence reagent kit for use with a suitably equipped flow cytometer to identify and determine the percentages and absolute counts of the following mature human lymphocyte subsets in erythrocyte-lysed whole blood: T lymphocytes (CD3+), B lymphocytes (CD19+), helper/inducer T lymphocytes (CD3+CD4+), suppressor/cytotoxic T lymphocytes (CD3+CD8+), and natural killer (NK) lymphocytes (CD3–CD16+and/or CD56+).
Catalog No. NC1700887
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.