Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD NIH 3T3 + Vanadate + PDGF Lysate

Catalog No. BDB612069 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612069 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612069 Supplier BD Supplier No. 612069

missing translation for 'validMainframeMsg'

Includes

Mouse Embryonic Fibroblast

Useful control for the western blotting application

  • Can be rich in endogenously expressed proteins
  • Convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel

TRUSTED_SUSTAINABILITY

Specifications

Tissue Embryonic Fibroblast
Preparation Method NIH 3T3 cells were Serum starved overnight and then incubated at room temperature for 10 min. in a final concentration of 1mM Sodium Vanadate, PDGF was then added at a final concentration of 5ng/mL and cells were incubated at 37°C
Quantity 500 μg
Storage Buffer 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Host Species Mouse
Concentration 1 mg/mL
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.