Learn More
Description
- Composed of extracellular domain of mouse CTLA-4 fused to mutant Fc region of mouse IgG2a
- Binds to CTLA-4's ligands B7-1 (CD80) and B7-2 (CD86) on mouse antigen-presenting cells
- Expressed on activated T lymphocytes 2-3 days after stimulation through T-cell receptor
- Prevent allograft rejection and induce donor-specific tolerance
- Storage buffer: Aqueous buffered solution containing no preservative
Blocking
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | Blocking Agent |
| Formulation | Aqueous Buffered solution containing no preservative |
| Name | Mouse NA/LE Non-Cytolytic CTLA-4 - IgG Fusion Protein |
| Quantity | 0.5 mg |
| Storage Requirements | -20°C |
| Cross Reactivity | Mouse |
| Classification | No Azide/Low Endotoxin (NA/LE) |
| Form | Lyophilized |
For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.