Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD OptEIA™ Assay Diluent

Catalog No. bdb555213 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555213 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555213 Supplier BD Supplier No. 555213
Only null left
Add to Cart
Add to Cart

A buffered protein solution containing fetal bovine serum that is intended for use in BD OptEIA ELISA Sets

  • Serves as a blocking agent for ELISA plates
  • Serves as a diluent for ELISA standards/samples
  • Used during the preparation of the Working Detector described in specific BD OptEIA ELISA Set protocols

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Immunoreagent Assay Diluent
For Use With (Application) ELISA
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.