Learn More
Description
Includes
Capture antibody, detection antibody, streptavidin-HRP standards.
- Size: 20 Plates
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 20 Plates |
| Species | Human |
| Cross Reactivity | Human |
| Form | Liquid |
| For Use With (Application) | ELISA |
| Kit Contents | Capture Antibody, detection Antibody, Streptavidin-HRP, standards |
| Content And Storage | 2°C to 8°C |
| Research Area | Cytokines |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.