Learn More
Description
Includes
Calcium Indicator, 10X PBX Signal Enhancer
- Involves one dye addition step
- Does not require washing
- Allows easy implementation in highthroughput environment
- Probenecid, a standard inhibitor of nonspecific anion transport, is not required for dye-loading
- Can improve retention of the dye in cells
Specifications
Specifications
| Content And Storage | Calcium indicator at -20°C and PBX Signal Enhancer 10X at room temperature |
| Detection Method | Fluorometric |
| For Use With (Application) | For homogeneous measurement of intracellular calcium changes |
| Includes | 10 vials calcium indicator,100mL PBX signal enhancer (10X, lyophilized) |
| Quantity | 100 Preps |
| Type | Calcium Assay Kit |
| Druggable Target | Calcium Signaling |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.