Learn More
Description
The TW4-9E7 monoclonal antibody specifically binds to Human Transforming Growth Factor beta-1 (TGF-β1). TGF-β1 is a potent multifunctional cytokine that positively or negatively regulates numerous processes including development, hematopoiesis, tissue remodeling, wound repair, innate and adaptive immunity as well as cancer and autoimmune diseases. TGF-β1 is formed by the enzymatic cleavage of the TGF-β1 propeptide that is encoded by the TGFβ1 gene and comprised of the Latency Associated Peptide (LAP) and TGF-β1. Prior to secretion, the dimeric LAP-TGF-β1 propeptide is cleaved resulting in a biologically inactive form of dimeric TGF-β1 that is noncovalently associated with dimeric LAP (latent TGF-β1). This complex may be expressed on the surface of TGF-β1-producing cells or be further processed by proteolytic removal of LAP to release the biologically active mature form of the soluble TGF-β1 homodimer. Many different cell types synthesize TGF-β1 and express specific receptors for it.
- Alternate name: TGFB1; TGFβ1; TGF-β-1; Transforming growth factor, β 1; CED; LAP
- Clone: TW4-9E7
- Immunogen: Human TGF-β1 transfected cell line
- Isotype: Mouse (BALB/c) IgG1, κ
- Strain host: BALB/c
- Reactivity: Human
- Format: PE
- Volume: 5μL
- Storage Buffer: Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Intracellular staining (flow cytometry), Flow cytometry
Specifications
Specifications
| For Use With (Application) | Flow Cytometry, Intracellular Staining |
| Formulation | Aqueous Buffered solution containing BSA and 0.09% Sodium Azide |
| Molecular Weight (g/mol) | 14.5 |
| Name | Mouse anti-Human TGF-beta1 |
| Quantity | 100 Tests |
| Source | BALB/c |
| Storage Requirements | 4°C |
| Conjugate | PE Labeled |
| Cross Reactivity | Mouse |
| Form | Liquid |
Research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.