Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Perm/Wash™ Buffer

Catalog No. bdb554723 Shop All BD Biosciences Products
Click to view available options
Quantity:
100 mL

Catalog No. BDB554723

Supplier: BD 554723

Only null left
Add to Cart
Add to Cart

Can be used in intracellular cytokine staining to permeabilize cells and as an antibody diluent and cell wash buffer

  • 10X stock solution containing both Fetal Bovine Serum (FBS) and saponin

It is not uncommon for the color of this product to vary from lot to lot. A small amount of insoluble precipitate is also common. Color variation and/or precipitate do not affect product performance.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Fetal Bovine Serum (FBS) Solution 10X and Saponin
For Use With (Application) Can be used in intracellular Cyto kine staining to permeabilize cells and as an Antibody diluent and cell Wash Buffer
pH Range 7.2 to 7.6
Content And Storage 4°C
Product Type Perm or Wash Buffer
Quantity 100 mL
Format Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.