Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Blue Live Cell Caspase Probe

Catalog No. BDB565519 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565519 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565519 Supplier BD Supplier No. 565519
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cells with active caspases can be detected. BD Pharmingen™ Blue Live Cell Caspase Probe contains a fluorochrome-labeled caspase inhibitor. Fluorochrome-labeled caspase inhibitors have three functional domains: fluorochrome , 3-amino acid sequence (VAD) that binds to the active site of the activated caspase, and fluoromethyl ketone (FMK) moiety that allows the probe to irreversibly bind the active caspase.

  • Blue Live Cell Caspase Probe is excited by the blue laser (with an excitation maximum of 492nm), and has a fluorescence emission maximum of 514nm
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 Tests
Reactivity Human, Mouse
Product Type Blue Live Cell Caspase Probe
Content And Storage -20°C
For Use With (Application) Flow Cytometry, Intracellular Staining(Flow Cytometry)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.