Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosflow Lyse/Fix Buffer 5X, BD

Catalog No. bdb558049 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558049 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558049 Supplier BD Biosciences Supplier No. 558049
Add to Cart
Add to Cart

For the lysing and fixing of whole blood for use with intracellular flow cytometry

  • Can be used for lysing red blood cells and for fixing leukocytes in one step, allowing investigators to rapidly inactivate kinases, phosphatases and/or proteases, so that the in vivo phosphorylation state of the cell can be examined
  • Has been reported to preserve the light scattering properties of the cell
  • May be used on human and mouse whole blood
  • Whole blood lysis has been shown to be as effective as density gradient centrifugation in the preparation of PBMCs for lymphocyte subset analysis
  • In many cases, phospho-specific antibodies may be used simultaneously with the staining of cell surface markers (e.g. CD markers); however, not all cell surface markers, however, are compatible with the BD Phosflow Lyse/Fix Buffer

TRUSTED_SUSTAINABILITY

Specifications

Product Type Lyse/Fix Buffer
Format Liquid
Validated Application RUO
Color Clear
Quantity 250mL
Content And Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.