Learn More
BD Phosflow™ Violet Fluorescent Cell Barcoding Kit
Catalog No. BDB561570 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
800 Tests
Catalog No. Quantity
BDB561570 800 Tests
1 options
Catalog No. BDB561570 Supplier BD Supplier No. 561570
Add to Cart
Add to Cart
Includes
Cell Barcoding Dye 450 (CBD450) and Cell Barcoding Dye 500 (CBD500), Fluorescent Cell Barcoding Wash Buffer, and choice of one Alexa Fluor™ 647-conjugated, phospho-specific antibody
Provides reagents for successfully barcoding T cell subsets from human whole blood samples for studying phosphorylated protein induction in MAP Kinase and JAK/STAT signaling pathways
  • Reactivity: human
  • Size: 800 tests
TRUSTED_SUSTAINABILITY
Specifications
Quantity 800 Tests
Reactivity Human
WARNING: Reproductive Harm - www.P65Warnings.ca.gov