Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Phospholipid-binding Protein, APC Annexin V

Catalog No. BDB550474 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
200 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550474 100 Tests
BDB550475 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB550474 Supplier BD Supplier No. 550474
Add to Cart
Add to Cart

Has high affinity for PS, and binds to cells with exposed PS

  • Conjugated to APC
  • Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Tests
Sample Volume 5 μL
Storage Buffer Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Conjugate APC (Allophycocyanin)
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.