Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Phospholipid-binding Protein, APC Annexin V
Click to view available options
Quantity:
100 Tests
200 Tests
Description
- Conjugated to APC
- Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes
Flow Cytometry

Specifications
Specifications
Quantity | 100 Tests |
Sample Volume | 5 μL |
Storage Buffer | Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide |
Content And Storage | 4°C |
For Use With (Application) | Flow Cytometry |
Conjugate | APC (Allophycocyanin) |
Form | Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction