Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Prepared Media: Trypticase Soy Broth

Catalog No. 14959802 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
4 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-959-802 4 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-959-802 Supplier BD Supplier No. 299919
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

  • Format: Dehydrated
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L
Packaging Type Bottle
Quantity 4 x 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.