Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Protein A: HRP

Catalog No. BDB610438 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610438 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610438 Supplier BD Supplier No. 610438
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For routine western blotting analysis

  • Format: HRP
  • Storage buffer: 1mg/mL in 50% glycerol, 20mM sodium phosphate pH 7.5, 150 mM NaCl

Western blot

TRUSTED_SUSTAINABILITY

Specifications

Product Type HRP Protein A
Quantity 1 mg
Concentration 1 mg/mL
For Use With (Application) Western Blot
Content And Storage 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.