Learn More
BD Ratiometric Calcium Assay Kit
Catalog No. BDB644244 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 Preps
100 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB644244 100 Preps
BDB644243 10 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB644244 Supplier BD Supplier No. 644244
Includes
Ratiometric Calcium Indicator, lyophilized 10X Signal Enhancer
Involves single dye addition step with no subsequent washing
  • For homogeneous measurement of intracellular calcium changes caused by activation of G-protein coupled receptors or calcium channels
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
For Use With (Application) Kit allows homogeneous measurement of intracellular calcium changes caused by activation of G-proteins coupled receptors or calcium channels
Includes 10 vials calcium indicator,100mL signal enhancer (10X, lyophilized)
Quantity 100 Preps
Type Detection Assay Kit