Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human IL-15

Catalog No. BDB560254 Shop All BD Biosciences Products
Change view
Click to view available options
Size: 5μg; Reactivity: human
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BDB560254 Size: 5μg; Reactivity: human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560254 Supplier BD Supplier No. 560254
Only null left
Add to Cart
Add to Cart

Lyophilized sterile-filtered aqueous buffered protein solution containing 1mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives

Interleukin-15 (IL-15) is a potent immunoregulatory cytokine produced by a variety of cell types including monocytes and epithelial cells. This T cell growth factor can enhance cytolytic activity of antigen-specific T cells and lymphokine-activated killer cells. Human IL-15 is an 11 kDa protein, as determined by SDS-PAGE analysis.

  • Reactivity: Human
  • Molecular weight: 11KDa
  • >95% pure
  • Endotoxin level: ≤0.1ng per μg of human IL-15
  • Storage buffer: Lyophilized in an aqueous buffered solution containing BSA

Functional assay, ELISA Standard

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA Standard, Functional Assay
Formulation Lyophilized in an aqueous Buffered solution containing BSA
Molecular Weight (g/mol) 11
Name Human IL-15 Recombinant Protein
Quantity 5 μg
Storage Requirements -20°C
Cross Reactivity Human
Recombinant Recombinant
Form Lyophilized

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.