Learn More
Description
Interleukin-10 (IL-10), originally known as Cytokine Synthesis Inhibitory Factor (CSIF), is an 18.5 kD protein that shares over 80% sequence homology with Epstein-Barr Virus protein BCRFI.
- Reactivity: Mouse
Bioassay, Enzyme Linked Immunosorbent Assay, BioAssay
Specifications
Specifications
| Concentration | 200μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard |
| Name | Mouse IL-10 Recombinant Protein |
| Quantity | 10 μg |
| Storage Requirements | -80°C |
| Recombinant | Recombinant |
| Form | Liquid |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.