Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Mouse Interleukin-13 (IL-13)

Catalog No. BDB554599 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554599 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554599 Supplier BD Supplier No. 554599
Add to Cart
Add to Cart

Determined by SDS-PAGE, and an absorbance assay based on the Beers- Lambert law

Frozen aqueous buffered solution containing 1.0mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives

  • Reactivity: Mouse
  • Storage Buffer: Frozen aqueous buffered solution containing BSA
  • Activity Assay: Measured by proliferation using TF-1 indicator cells
  • Specific Activity Range: 0.6 - 5 x 107 Units/mg
  • ED50: 0.2 to 1.5ng/ml ; Observed dose response relationship: 50 fold

Bioassay

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100μg/mL
For Use With (Application) Bioassay
Formulation Frozen aqueous Buffered solution containing BSA
Name Mouse IL-13 Recombinant Protein
Purification Method SDS-PAGE
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Mouse
Recombinant Recombinant
Form Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.