Learn More
Description
Quantitative standard for measuring mouse IL-2 protein levels in an IL-2 specific sandwich ELISA with the purified JES6-1A12 antibody as capture antibody and the biotinylated JES6-5H4 antibody as detection antibody
- Storage buffer: Frozen aqueous buffered solution containing BSA
- Biological Activity: Activity Range is measured in a proliferation assay using CTLL-2 indicator cells
- Specific Activity: 0.1 - 1.0 x 1099 Units™/mg
- ED50: 10 - 100 pg/ml
Bioassay, Enzyme-linked immunosorbent assay standard
Specifications
Specifications
| Concentration | 200μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard |
| Formulation | Frozen aqueous Buffered solution containing BSA |
| Molecular Weight (g/mol) | 17.2 |
| Name | Mouse IL-2 Recombinant Protein |
| Quantity | 20 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Mouse |
| Recombinant | Recombinant |
| Form | Solution |
Research Use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.