Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Mouse Interleukin-3 (IL-3)

Catalog No. BDB554579 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554579 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554579 Supplier BD Supplier No. 554579
Add to Cart
Add to Cart

Species-specific cytokine which acts on hematopoietic progenitors of every lineage, with the probable exception of cells committed to the lymphoid lineage

Interleukin-3 (IL-3) has been referred to as CSF-2 , BPA, megakaryocyte CSF, HCGF and hemopoietin 2.

  • Molecular weight: 17.2 kDa
  • Contains 140 amino acid residues
  • Storage buffer: Frozen aqueous buffered solution containing BSA and glycerol
  • >95% pure
  • No preservatives

Bioassay, ELISA Standard, Blocking

TRUSTED_SUSTAINABILITY

Specifications

Concentration 200μg/mL
For Use With (Application) Bioassay, Blocking Agent, ELISA Standard
Formulation Frozen aqueous Buffered solution containing BSA and Glycerol
Molecular Weight (g/mol) 17.2
Name Mouse IL-3 Recombinant Protein
Quantity 10 μg
Storage Requirements -80°C
Cross Reactivity Mouse
Recombinant Recombinant
Form Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.