Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Mouse Interleukin-5 (IL-5)

Catalog No. BDB554581 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554581 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554581 Supplier BD Supplier No. 554581
Add to Cart
Add to Cart

Lymphokine for activities related to eosinophil differentiating factor, B cell growth factor II, T cell-replacing factor

  • 35-37kD homodimer
  • Induces eosinophil differentiation
  • Promotes survival and activation of eosinophil
  • Stimulates generation of cytotoxic T cell
  • Supports growth and differentiation of pre-activated B cells and hematopoietic cells
  • Reactivity: mouse
  • Storage buffer: frozen aqueous buffered solution containing BSA and glycerol

ELISA standard, functional assay, blocking

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100μg/mL
For Use With (Application) Blocking Agent, ELISA Standard, Functional Assay
Formulation Frozen aqueous Buffered solution containing BSA and Glycerol
Molecular Weight (g/mol) 35 to 37
Name Mouse IL-5 Recombinant Protein
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Mouse
Recombinant Recombinant
Form Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.