Learn More
Description
Supplied as a frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution, and containing 2mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
- Reavtivity: Rat
- Storage buffer: Frozen aqueous buffered solution containing BSA
- Activity Test: Biological activity is measured in an anti-viral bioassay using L929 indicator cells
- Specific Activity Range: 0.1-1.0x108Unit/mg
- ED50: 0.1 - 1.0 ng/ml
- Observed dose response relationship: 100 fold
Bioassay
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | Bioassay |
| Formulation | Frozen aqueous Buffered solution containing BSA |
| Name | Rat IFN-gamma Recombinant Protein |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Rat |
| Recombinant | Recombinant |
| Form | Solution |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.