Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Rat Interleukin-2 (IL-2)

Catalog No. BDB555106 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555106 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555106 Supplier BD Supplier No. 555106
Add to Cart
Add to Cart

Potent, multifunctional cytokine produced by activated T lymphocytes

  • Responsible for T cell growth and replacing factor, thymocyte stimulation and mitogenesis factor, killer helper factor
  • Stimulates growth and differentiation of T cells and cytokine production by T cells
  • Activates monocytes/macrophages and NK and LAK cells
  • Can augment B cell growth and differentiation
  • Reactivity: Rat
  • Storage buffer: frozen aqueous buffered solution containing BSA and glycerol

Bioassay, ELISA standard

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100μg/mL
For Use With (Application) Bioassay, ELISA Standard
Formulation Frozen aqueous Buffered solution containing BSA and Glycerol
Molecular Weight (g/mol) 16.5
Name Rat IL-2 Recombinant Protein
pH Range 7.2
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Rat
Recombinant Recombinant
Form Solution
Show More Show Less

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.