Learn More
BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) with Lecithin and Polysorbate 80 (LP80)
Shop All BD Diagnostic Systems ProductsDescription
The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.
The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:
- Withstands an accidental drop from the height of a bench-top workspace
- Ensures the integrity of the sample
- Prevents the dish from opening unintentionally
- Protects media from accidental cross contamination
- Improves stability during stacking and storage
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Additive | Lecithin, Polysorbate 80 |
| Form | Liquid |
| Format | Plate, Isolator Pack |
| Manufacturing Origin | United States |
| Media Type | TSA w/Lecithin, Polysorbate 80 |
| Product Type | Contact Plate |
| Sterility | Sterile |
| Target Organism Class | Fastidious Organisms |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.