Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Streptavidin Particles Plus - DM

Catalog No. BDB557812 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557812 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557812 Supplier BD Supplier No. 557812
Add to Cart
Add to Cart

For positive selection or depletion of leukocyte subpopulations using the BD IMagnet™

The small size of BD IMag particles allows the positive fraction to be further evaluated in downstream applications.

  • Magnetic nanoparticles
  • Have streptavidin covalently conjugated to their surface
  • Allow positive fraction to be further evaluated in downstream applications such as flow cytometry
  • Storage Buffer: Aqueous buffered solution containing BSA and ≤0.09% sodium azide

Cell separation

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mL
Content And Storage 4°C
For Use With (Application) Cell separation, for positive selection or depletion of leukocyte subpopulations using the BD IMagnet

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.