Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD SurePrep™ Capillary Tubes
Click to view available options
Additive:
Heparin
Heparin (Precalibrated)
Plain
Quantity:
10 Vials of 100 Tubes Each
2 Vials of 100 Tubes Each
Description
- Tubes feature self-sealing plug and multilayered Mylar™ wrap
- Reduce exposure to HIV, HBV, and other bloodborne pathogens
- A channel in the system allows air to escape during filling; once the sample touches the plug, the channel seals automatically
- Simply fill the tube, hold vertically, then centrifuge and read
- In the event of accidental breakage, the protective Mylar wrap helps contain blood and minimizes exposure to glass fragments

Specifications
Specifications
Color | Clear |
Additive | Heparin |
Material | Mylar Wrapped Glass |
Sterility | Non-sterile |
Closure Type | Self-sealing Plug |
Quantity | 10 Vials of 100 Tubes Each |
For Use With (Application) | Capillary Blood Collection Tube |
Description | Size: 1.1 X 75 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction