Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Transduction Laboratories Mouse Spleen Lysate

Catalog No. BDB611462 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611462 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Do not dilute or boil
Catalog No. BDB611462 Supplier BD Supplier No. 611462

missing translation for 'validMainframeMsg'

Do not dilute or boil

Ready to use for immediate loading onto an SDS-PAGE gel

  • For use as a control for western blotting
  • Rich source of endogenously expressed proteins
  • Provided in a denatured format

TRUSTED_SUSTAINABILITY

Specifications

Tissue Spleen
Quantity 500 μg
Host Species Mouse
Concentration 1 mg/mL
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.