Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Conventional Stopper

Catalog No. 23021018 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Diameter (Metric) Outer:
13 x 75 mm
13 x 100 mm
16 x 100 mm
Capacity (Metric):
3 mL
6 mL
10 mL
Product Type:
Barcoded Blood Collection Tube
Blood Collection Tube
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Capacity (Metric) Product Type
23-021-018 16 x 100 mm 10 mL Barcoded Blood Collection Tube
02-657-27 13 x 75 mm 3 mL Blood Collection Tube
02-685-112 16 x 100 mm 10 mL Blood Collection Tube
23-021-012 13 x 100 mm 6 mL Blood Collection Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-021-018 Supplier BD Supplier No. 368045

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Use for serum determinations in chemistry, serology and immunohematology.

Bar coded
TRUSTED_SUSTAINABILITY

Specifications

Product Type Barcoded Blood Collection Tube
Additive Clot Activator, Silicone Coated
Material Plastic
Closure Type Traditional Cap
Capacity (Metric) 10 mL
Closure Color Red
Diameter (Metric) Outer 16 x 100 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.