Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Violet Proliferation Dye 450

Catalog No. BDB562158 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562158 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562158 Supplier BD Supplier No. 562158
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For monitoring of cell divisions completed by single cells, intracellular staining

Flow Cytometry, Intracellular staining

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Content And Storage -20°C
For Use With (Application) Intracellular Staining, Monitoring Cell Divisions
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.