Learn More
CPC Scientific H-Arg-Thr-Arg-Pro-Leu-Trp-Val-Arg-Met-Glu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PANP004A
Encompass
The peptide was used in type 1 diabetes (T1D) study. BDC2.5 mimotope was utilized to support the study on antigen presentation of antigenic peptides to islet autoantigen-specific T cells.ONE-LETTER SEQUENCE: RTRPLWVRMEMOLECULAR FORMULA: C59H98N20O14S1MOLECULAR WEIGHT:1343.61STORAGE CONDITIONS: -20 5CRESEARCH AREA: GastrointestinalREFERENCES: Hu, C. et al. J. Clin. Invest. 117, 3857 (2007); Martin-Orozco, N. et al. J. Immunol. 171, 6954 (2003)
Catalog No. 50-211-0080
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.