Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Veal Infusion Broth

Catalog No. DF0344176 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0344-17-6 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0344-17-6 Supplier BD Supplier No. 234420
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivating fastidious microorganisms

  • Formulation: Lean Veal infusion from 500g 10g/L, No. 3 Proteose Peptone 10g/L, Sodium Chloride 5g/L

Order Info

Prices are for units of one each unless otherwise specified

Certifications

AOAC, BAM, COMPF

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, COMPF
Content And Storage <30°C
Form Dehydrated
Media Type Veal Infusion Broth
Product Type Microbiology Media
Formulation Lean Veal infusion from 500g 10g/L, No. 3 Proteose Peptone 10g/L, Sodium Chloride 5g/L
For Use With (Application) Cultivation of fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.