Learn More
Stemculture LLC StemCultures LLC BDNF DISCs (Standard) - 5 ng/mL in 2 mL
SDP
Supplier:  Stemculture LLC DSCBD112
Encompass_Preferred
Note: For 2D cultures, place DISC in a transwell to avoid cell lifting. For 3D cultures (i.e. organoids) place DISC directly in the culture well. BDNF DISCs are patented non-degradable, biocompatible hydrogels that release defined levels of native BDNF stably into culture medium over two weeks. BDNF DISCs are loaded with StemBeads BDNF, PLGA microbeads that encapsulate the protein. BDNF is key for supporting neuronal differentiation. BDNF DISCs are easy to add and remove, giving scientists enhanced control of growth factor levels in their cultures. Controlled delivery and stable levels overcome the 2 hour half-life of BDNF and improve cell cultures while saving researchers valuable time and resources, allowing for drastically improved neuron quality. BDNF DISCs release at 5 ng/mL in 2 mL of medium. BDNF DISCs have been tested in multiple neuronal base media, including Brain Phys, with enhanced cellular profiles. In these cases, BDNF DISCs were used with and without GDNF DISCs.
Catalog No. NC2670046
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.