Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech BDNF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. p-7245121
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-8442 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-8442 Supplier PeproTech Supplier No. 500P84BT500UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MHSDPARRGE LSVCDSISEW VTAADKKTAV DMSGGTVTVL EKVPVSKGQL KQYFYETKCN PMGYTKEGCR GIDKRHWNSQ CRTTQSYVRA LTMDSKKRIG WRFIRIDTSC VCTLTIKRGR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human/Murine/Rat BDNF. Anti-Human/Murine/Rat BDNF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human BDNF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BDNF (500-P84) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BDNF. Western Blot: To detect hBDNF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBDNF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P84BT-1MG will be provided as 2 x 500 μg

BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer's and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BDNF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene BDNF
Gene Accession No. P21237, P23363, P23560
Gene Alias abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF
Gene Symbols BDNF
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human/Murine/Rat BDNF.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12064, 24225, 627
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.