Learn More
Scientific Industries Bead Genie, 120V Type G plug, UK
Supplier:  Scientific Industries SIB103
Encompass_Preferred
Bead Genie, 120V Type G plug, UK. Bead beater ideal for lysing, extraction, or homogenization applications, utilizing an aggressive vertical mixing motion. Supplied with holders to hold vertically 12 microtubes (1.5 or 2.0ml), 6 15ml tubes, or 3 50ml tubes, and 12 microtubes (1.5 or 2.0ml) horizontally. Beads available and sold separately for complete bead beating solution. The Bead Genie, built in the USA, is compact compared to most bead beaters and homogenizers.
Catalog No. 50-206-7595
Only null left
Add to Cart
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.