Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp Scientifica™ 400 ml Beaker
SDP

Catalog No. 11392157
Change view
Click to view available options
Capacity (Metric):
400 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
11-392-157 400 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-157 Supplier Velp Scientifica™ Supplier No. A00000999
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Velp Scientifica™ 400 ml Beaker for use with Enzymatic Digester GDE.

  • For use with Enzymatic Digester GDE

Specifications

Product Type Beaker
Capacity (Metric) 400 mL
For Use With (Equipment) Enzymatic Digester GDE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.