Learn More
Andwin Scientific BEAKER W/HANDLE 2000ML
Supplier: Andwin Scientific 13916762
The 2 L Graduated Beaker with Handle are sturdy beakers made of autoclavable, translucent polypropylene. Its low form structure provides better stability. This product is graduated in the range of 502000 mL, which allows precise sample measurement.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.